Would you like to log in again?
Bürkle Cylindrical Barbed Y-Connectors, PVDF
Supplier: BURKLE INC
8712-0006 8712-0004 8712-0012 8712-0014 8712-0010 8712-0008
MFLX06044-19EA 32.5 USD InStock
MFLX06044-19 MFLX06044-21 MFLX06044-16 MFLX06044-15 MFLX06044-18 MFLX06044-17
Bürkle Cylindrical Barbed Y-Connectors, PVDF
Fittings and Connectors
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The cylindrical barbed Y connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty