Would you like to log in again?
Cylindrical Barbed 90º Elbow Connectors, PVDF
Supplier: BURKLE INC
8713-0004 8713-0010 8713-0014 8713-0006 8713-0012 8713-0008
MFLX06044-23EA 11.2 USD InStock
MFLX06044-23 MFLX06044-22 MFLX06044-25 MFLX06044-24 MFLX06044-27 MFLX06044-26
Cylindrical Barbed 90º Elbow Connectors, PVDF
Fittings and Connectors
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The cylindrical barbed 90º elbow connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty