Would you like to log in again?
Cylindrical Barbed Cross Connectors, PVDF
Supplier: BURKLE INC
8717-0008 8717-0006 8717-0012 8717-0010 8717-0004 8717-0014
MFLX06044-32EA 30.3 USD InStock
MFLX06044-32 MFLX06044-31 MFLX06044-34 MFLX06044-33 MFLX06044-29 MFLX06044-28
Cylindrical Barbed Cross Connectors, PVDF
Fittings and Connectors
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The cylindrical barbed cross connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty