Would you like to log in again?
Union Fittings, Hose Barb, Y-Connector, Polyproylene, Dynalon®
Supplier: Dynalon Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
222585 100PK 227025 100PK 222555 100PK 222575 100PK 222595 100PK 222565 100PK 222605 100PK 222545 100PK 227045 100PK 227085 100PK
MFLX06295-10BLKEA 72.7 USD InStock
MFLX06295-10BLK MFLX06295-60BLK MFLX06295-70BLK MFLX06296-20BLK MFLX06296-00BLK MFLX06295-40BLK MFLX06296-50BLK MFLX06295-50BLK MFLX06295-30BLK MFLX06295-20BLK
Union Fittings, Hose Barb, Y-Connector, Polyproylene, Dynalon®
Fittings and Connectors
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty