Would you like to log in again?
CPC® APC Acetal Quick-Disconnect Fittings, Bodies and Inserts
Supplier: CPC (Colder Products Company)
APCD23006 APC17006SH APCD17004SH APCD24004BSPT APCD16006 APCD17006SH APC24004BSPT APC17006SH 25PK APCD23006 25PK APCD24006 APCD16004 APCD12006 APC17004SH APC16004 APCD16005 APCD24004BSPT 25PK APC200M10 APC16006 25PK APCD10004 25PK APC12004 APCD10004 APCD10006 APCD12004 APC16006 APC17004SH 25PK
MFLX06353-01BLKEA 149 USD InStock
MFLX06353-01BLK MFLX49007-36 MFLX06353-19 MFLX06353-08BLK MFLX06353-17 MFLX06353-15 MFLX06353-34 MFLX06353-33 MFLX06353-11 MFLX06353-21BLK MFLX06353-25BLK MFLX06353-09 MFLX06353-34BLK MFLX06353-07 MFLX06353-08 MFLX06353-28 MFLX06353-07BLK MFLX06353-25 MFLX06353-26 MFLX06353-23 MFLX06353-01 MFLX06353-24 MFLX06353-02 MFLX06353-21 MFLX06353-22
CPC® APC Acetal Quick-Disconnect Fittings, Bodies and Inserts
Fittings and Connectors
Fittings connect in a snap.
  • No exposed metal parts
  • Protects against accidental disconnects
These CPC® APC-Series quick-disconnect fittings, combine a compact design with durable acetal construction, making them an excellent choice for many applications. All fittings feature a 1/4” flow, plastic thumb latch, fewer moving parts, and a smooth contoured design to deliver ease-of-use and excellent flow in a compact size. Simply press the thumb latch to disconnect fitting halves. Choose from in-line compression and hose barb bodies which feature shrouded latches to provide further protection against accidental disconnect. The ferruleless PTF compression fittings do not require ferrules to achieve a secure connection, which makes them easier to use and reuse. PTF fittings are designed for semi-rigid tubing, i.e., polyethylene, nylon, polyurethane, etc.
A complete coupler requires a fitting body and an insert (order separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty