Would you like to log in again?
CPC® Quick-Disconnect Fittings, Threaded Bodies
Supplier: CPC (Colder Products Company)
PMCD1004 (25PK) PMC1002 (25PK) PMCD1002 (1/PK) PMC1004 PLCD10006 (25PK) PLCD1000612 (1/PK) PLC10006 PMCD1004 (1/PK) PLCD1000612 (25PK) PMCD100412 (1/PK) PLCD10004 (1/PK) PLC10004 PMCD100212 (1/PK) PLCD10006 (1/PK) PLCD10004 (25PK) PLCD1000412 (1/PK) PLCD1000412 (25PK) PMCD100212 (25PK) PMCD100412 (25PK) PMCD1002 (25PK) PMC1002 (1/PK)
MFLX06359-71BLKEA 193 USD InStock
MFLX06359-71BLK MFLX06361-01 MFLX06361-00 MFLX06361-43 MFLX06359-70BLK MFLX06359-73BLK MFLX06361-42 MFLX06359-72BLK MFLX06361-00BLK MFLX06359-83BLK MFLX06359-82BLK MFLX06359-81BLK MFLX06359-80 MFLX06359-70 MFLX06359-81 MFLX06359-71 MFLX06359-82 MFLX06359-80BLK MFLX06359-83 MFLX06359-72 MFLX06359-73
CPC® Quick-Disconnect Fittings, Threaded Bodies
Fittings and Connectors
No more struggling—just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty