Would you like to log in again?
CPC® Plastic Quick-Disconnect Fittings, Elbow Compression Inserts
Supplier: CPC (Colder Products Company)
APC21004 25PK APC21006 APC21004
MFLX06360-31EA 252.4 USD InStock
MFLX06360-31 MFLX06360-34 MFLX06360-31BLK
CPC® Plastic Quick-Disconnect Fittings, Elbow Compression Inserts
Fittings and Connectors
Fittings connect in a snap.
  • Constructed of durable acetal plastic—ideal for use with corrosive fluids
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
A complete coupler requires a fitting body and an insert (order separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty