Would you like to log in again?
CPC® Quick-Disconnect Fittings, Hose Barb Elbow Inserts
Supplier: CPC (Colder Products Company)
PMCD230412 PMCD230412 (25PK) PMCD2302 (25PK) PLC2300412 (1/PK) PMC230212 PLC23006 PMCD2304 (25PK) PLC23004 (25PK) PLCD2300412 (25PK) PLCD23004 (25PK) PLCD2300412 PMCD230212 (25PK) PMCD2304 (1/PK) PMCD230212 (1/PK) PMC2304 (25PK) PMC230412 PMC2302 (1/PK) PLCD2300612 PLCD23006 (1/PK) PLCD23006 (25PK) PLC2300412 (25PK) PLC23004 (1/PK) PLCD23004 (1/PK) PLC23006 PMC230412 PLC2300612 PMCD2302 (1/PK) PMC2304 (1/PK) PLC2300612 PLCD2300612 (25PK) PMC2302 (25PK)
MFLX06362-00BLKEA 316 USD InStock
MFLX06362-00BLK MFLX06361-99BLK MFLX06361-87 MFLX06361-28BLK MFLX06361-64 MFLX06361-27 MFLX31307-68 MFLX31307-67 MFLX06361-85 MFLX06361-41 MFLX06361-63 MFLX31307-67BLK MFLX06361-76BLK MFLX06361-39BLK MFLX06361-64BLK MFLX06361-41BLK MFLX06361-28 MFLX06361-87BLK MFLX06361-99 MFLX06362-01 MFLX06362-00 MFLX06361-98 MFLX06361-76 MFLX06361-75 MFLX06361-27BLK MFLX06361-98BLK MFLX06361-75BLK MFLX31307-68BLK MFLX06362-01BLK MFLX06361-63BLK MFLX06361-39
CPC® Quick-Disconnect Fittings, Hose Barb Elbow Inserts
Fittings and Connectors
No more struggling—just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty