Would you like to log in again?
CPC® Miniature Quick-Disconnect Fittings, Panel-Mount Hose Barb Inserts
Supplier: CPC (Colder Products Company)
SMMPMM3 SMMPM01 50PK SMMPM02 SMMPM01 SMMPMM3 50PK SMMPM0112 50PK SMMPM0212 SMMPM0112
MFLX06363-70BLKEA 203 USD InStock
MFLX06363-70BLK MFLX06363-80 MFLX06363-70 MFLX06363-71BLK MFLX06363-71 MFLX06363-80BLK MFLX06363-82 MFLX06363-72
CPC® Miniature Quick-Disconnect Fittings, Panel-Mount Hose Barb Inserts
Fittings and Connectors
No more kinked tubing—allows tubing to rotate freely.
  • Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
  • Tubing can rotate freely when connected—preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle 1/16" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.
For a complete coupling, order one fitting body and one insert.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty