Would you like to log in again?
CPC® Quick-Disconnect Fittings, Hose Barb Inserts
Supplier: CPC (Colder Products Company)
PMCD2203 (1/PK) PMC220412 PMC4204 PLC4200412 25PK PMCD4202 (1/PK) PMCD2201 (1/PK) PMCD4202 (25PK) PLC22006 (25PK) PLCD4200612 PMC2201 (1/PK) PMC220212 PMC220212 (25PK) PMCD2204 (1/PK) PLCD22006 (1/PK) PMCD220412 PLCD42004 PLCD2200612 PLCD22005 (25PK) PLCD4200412 PLCD4200412 25PK PMCD220112 (25PK) PMC4202 25PK PMC2202 (1/PK) PMCD220212 (25PK) PLCD22004 (1/PK) PMCD2201 (25PK) PLCD42004 (25PK) PMCD2202 (1/PK) PMC2202 (25PK) PMC2203 (1/PK) PLCD2200412 (25PK) PMCD2203 (25PK) PMC2204 PLCD22005 (1/PK) PMC4201 PMC220112 (25PK) PLC22005 PLC22004 (1/PK) PMC4203 PLCD22004 (25PK) PMC4202 PLCD2200412 PMCD220112 PLC2200612 PLC2200412 (25PK) PMC220112 PMC4201 25PK PLC22004 (25PK) PLCD42005 PLCD42006 PLC42006 PLCD42005 25PK PLC42004 PMCD2202 (25PK) PMCD2204 (25PK) PMC2201 (25PK) PMC2203 (25PK) PMCD220212 PLCD2200612 (25PK) PLC2200412 PLC22006 (1/PK) PMCD220412 (25PK) PMCD4203 PMC220412 (25PK) PLCD22006 (25PK) PLC4200412
MFLX31307-69BLKEA 319 USD InStock
MFLX31307-69BLK MFLX06361-23 MFLX06364-42BLK MFLX31307-09 MFLX06360-85BLK MFLX06361-61 MFLX31307-61BLK MFLX31307-14BLK MFLX06360-42BLK MFLX06360-43 MFLX06360-87 MFLX06360-44 MFLX06360-82BLK MFLX06360-85 MFLX06360-42 MFLX06364-45 MFLX06360-47 MFLX06364-47 MFLX06360-45 MFLX06361-58 MFLX06364-45BLK MFLX06364-39BLK MFLX06364-85 MFLX06364-42 MFLX06360-80 MFLX06364-87 MFLX06360-38BLK MFLX06360-83 MFLX06360-40 MFLX06360-84 MFLX06364-82 MFLX06361-93 MFLX06360-82 MFLX06364-40 MFLX06360-45BLK MFLX06360-87BLK MFLX06364-80BLK MFLX06364-38 MFLX06364-39 MFLX06364-40BLK MFLX06360-36 MFLX31307-69 MFLX06360-83BLK MFLX06364-38BLK MFLX06361-34BLK MFLX31307-64 MFLX31307-61 MFLX31307-62 MFLX06360-44BLK MFLX06364-80 MFLX06364-47BLK MFLX06360-38 MFLX06360-40BLK MFLX06361-34 MFLX06360-80BLK MFLX31307-19 MFLX31307-14 MFLX06364-85BLK MFLX06361-35 MFLX06361-71BLK MFLX06360-36BLK MFLX31307-64BLK MFLX06361-71 MFLX31307-09BLK MFLX06360-43BLK MFLX06364-82BLK
CPC® Quick-Disconnect Fittings, Hose Barb Inserts
Fittings and Connectors
No more struggling—just press thumb latch.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Fittings connect in a snap
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. The audible click when the coupling engages confirms a solid connection. To disconnect fitting halves, simply press the button on the coupling body. Standard flow quick-disconnect couplings require a body and an insert of the same flow size.
A complete coupler requires a fitting body and an insert (order separately).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty