Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Transfer Tubing, Perfluoroalkoxy (PFA), Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
97331 97387 39777 97386 43410 49433 97336 39780 97329 43406 39775 97390 43370 97330 91162 302180 44615 42315 40257 68940 48601 56202 97333
MFLX06371-28EA 460 USD InStock
MFLX06371-28 MFLX06371-06 MFLX06371-05 MFLX06371-08 MFLX06371-29 MFLX06371-07 MFLX06375-04 MFLX06371-22 MFLX06371-00 MFLX06375-02 MFLX06371-21 MFLX06375-01 MFLX06371-02 MFLX06375-08 MFLX06371-24 MFLX06371-23 MFLX06371-01 MFLX06375-07 MFLX06371-04 MFLX06371-26 MFLX06371-25 MFLX06375-05 MFLX06371-03
Masterflex® Transfer Tubing, Perfluoroalkoxy (PFA), Avantor®
Tubing Transfer Tubing
Stands up to highly corrosive materials.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application

Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. For the transfer of high-temperature or highly corrosive materials. Also good for high-purity work.
Characteristics: Chemically inert, nontoxic, and virtually nonporous. Excellent crack- and stress-resistance. Low coefficient of friction. Translucent.
Resin certification: Resin is FDA-compliant (21 CFR 177.1550); meets USP Class VI requirements. Tubing is manufactured according to GMP.
Temperature range: -320 to 500 °F (-195 to 260 °C)
Sterilize: Autoclave, ethylene oxide, or use dry heat.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty