Would you like to log in again?
Masterflex® Transfer Tubing, Vinyl, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
ABH00027-CP ABH00007-CP ABH00002-CP ABH00012-CP ABH00053-CP ABH00017-CP ABH02023-CP ABH00038-CP ABH00022-CP ABH02037-CP ABH02028-CP ABH00046-CP ABH02014-CP ABH00019-CP ABH00062-CP ABH02029-CP ABH02018-CP ABH02045-CP ABH00071-CP ABH00036-CP
MFLX06405-09EA 45 USD InStock
MFLX06405-09 MFLX06405-06 MFLX06405-27 MFLX06405-16 MFLX06405-19 MFLX06405-08 MFLX06405-07 MFLX06405-18 MFLX06405-02 MFLX06405-46 MFLX06405-12 MFLX06405-01 MFLX06405-15 MFLX06405-04 MFLX06405-03 MFLX06405-36 MFLX06405-25 MFLX06405-31 MFLX06405-20 MFLX06405-10
Masterflex® Transfer Tubing, Vinyl, Avantor®
Tubing Transfer Tubing
Ideal for general-purpose applications.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
The tubing is ideal for general-purpose lab use. Characteristics include fair chemical and abrasion resistance, odorless, nontoxic, and transparent. Tubing is manufactured according to GMP and has a temperature range of -45 to 180 °F (-43 to 82 °C). Can be sterilized with Ethylene oxide.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty