Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Transfer Tubing, Peroxide-Cured Silicone, Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
ABX00028 ABX00002 100270MM 100289MM ABX00038 ABX00014 ABX00027 100123MM 100017MM 100297MM ABX00023 ABX00046 100231MM 100280MM ABX00018 ABX00022 ABX00053 100303MM 100282MM ABX00017 100227MM ABX00019 100279MM 100124MM 100281MM 100133MM 100283MM 100122MM 100287MM 100019MM ABX00004 ABX00001 ABX00006 ABX00009 100309MM 100295MM ABX00007 ABX00024 ABX00016 ABX00012 ABX00011 100291MM 100299MM 100307MM 100301MM ABX00036 100278MM 100305MM ABX00029 100293MM
MFLX06411-03EA 156 USD InStock
MFLX06411-03 MFLX06411-60 MFLX95886-06 MFLX95886-07 MFLX95886-04 MFLX95886-05 MFLX95886-02 MFLX95886-03 MFLX95886-00 MFLX95886-01 MFLX06411-59 MFLX06411-19 MFLX06411-12 MFLX06411-55 MFLX06411-58 MFLX95886-10 MFLX06411-70 MFLX06411-72 MFLX06411-71 MFLX95886-17 MFLX95886-18 MFLX95886-15 MFLX95886-16 MFLX95886-13 MFLX95886-14 MFLX95886-11 MFLX95886-12 MFLX95886-08 MFLX95886-09 MFLX06411-63 MFLX06411-62 MFLX06411-65 MFLX06411-64 MFLX06411-67 MFLX06411-66 MFLX06411-69 MFLX06411-68 MFLX95886-21 MFLX06411-80 MFLX06411-83 MFLX06411-82 MFLX95886-24 MFLX95886-23 MFLX95886-19 MFLX06411-74 MFLX06411-76 MFLX06411-75 MFLX06411-78 MFLX06411-77 MFLX06411-79
Masterflex® Transfer Tubing, Peroxide-Cured Silicone, Avantor®
Tubing Transfer Tubing
No need to worry about the heat - use for applications up to 460 °F.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application

Applications: Pharmaceutical, biotechnology, food and beverage, aerospace, and general lab applications
Characteristics: Peroxide-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent
Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI requirements. Tubing is manufactured according to GMP.
Temperature range: -60 to 460 °F (-51 to 238 °C)
Sterilize: Autoclave or gamma irradiation

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty