Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Transfer Tubing, Tygon® ND-100-80 Microbore, Non-DEHP, Avantor®

Supplier: Avantor Fluid Handling
Masterflex®
AAD04103-CP ND-100-80 AAD04127-CP ND-100-80 AAD04119-CP ND-100-80 AAD04091-CP ND-100-80 AAD04133-CP ND-100-80
MFLX06420-01EA 426 USD InStock
MFLX06420-01 MFLX06420-05 MFLX06420-04 MFLX06420-03 MFLX06420-02
Masterflex® Transfer Tubing, Tygon® ND-100-80 Microbore, Non-DEHP, Avantor®
Tubing Transfer Tubing
High resistance to kinking & collapse.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application

Formulation: Tygon ND-100-80
Applications: General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.
Characteristics: Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear.
Certifications: Resin meets USP Class VI requirements, non-DEHP.
Temperature range: -31 to 185 °F (-35 to 85 °C)
Sterilize: Autoclave, ethylene oxide, or gamma irradiation.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty