Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Viton® Transfer Pump Tubing

Supplier: VWR International
T6434-02 T6434-09 T6434-05 T6434-07 T6434-08 T6434-04 T6434-03 T6434-06 T6434-01
MFLX06434-04CS 106 USD InStock
MFLX06434-04 MFLX06434-03 MFLX06434-02 MFLX06434-01 MFLX06434-09 MFLX06434-08 MFLX06434-07 MFLX06434-06 MFLX06434-05
Viton® Transfer Pump Tubing
Tubing Transfer Tubing
keep light sensitive samples safe.

  • Global market leader in range of formulations for most extensive chemical compatibility

Applications: Acid and solvent transfer; high-temperature work.
Characteristics: Opaque black.
Certification: Tubing is manufactured according to GMP.
Temperature range: –25 to 399 °F (–32 to 204 °C)
Sterilize: Not recommended.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty