Would you like to log in again?
Masterflex® Transfer Tubing, Fluorinated Etyhlene Propylene (FEP), Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
53281 50389 50655 14311 59369 14315 50694 43320 59973 73009 44263 53285 49438 52851 54072 44517 50627 49788 49849 54849 49845 81413 43259
MFLX06450-19EA 491 USD InStock
MFLX06450-19 MFLX06450-08 MFLX06450-07 MFLX06450-09 MFLX06406-18 MFLX06406-76 MFLX06406-64 MFLX06406-12 MFLX06406-66 MFLX06406-72 MFLX06406-60 MFLX06406-74 MFLX06406-63 MFLX06406-62 MFLX06450-00 MFLX06450-02 MFLX06450-01 MFLX06450-15 MFLX06450-04 MFLX06450-14 MFLX06450-06 MFLX06450-17 MFLX06450-05
Masterflex® Transfer Tubing, Fluorinated Etyhlene Propylene (FEP), Avantor®
Tubing Transfer Tubing
Does not deteriorate with age.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Excellent for the transfer of extreme temperature or highly corrosive materials. Also good for high-purity work.
Characteristics: Chemically inert, nontoxic, heat resistant, with a low coefficient of friction. Doesn’t deteriorate with age. Tough, but bends even at -454 °F (-270 °C). Translucent.
Certifications: Resin is FDA-compliant (21 CFR 177.1550). Tubing is manufactured in ISO 9001-2008 approved facility.
Temperature range: -454 to 400 °F (-270 to 205 °C)
Sterilize: Autoclave, ethylene oxide, or use dry heat.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty