Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® Transfer Tubing, Fluorinated Etyhlene Propylene (FEP), Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
53281 50389 50655 14311 59369 14315 50694 43320 59973 73009 44263 53285 49438 52851 44517 54072 50627 49788 49849 54849 49845 81413 43259
MFLX06450-19EA 491 USD InStock
MFLX06450-19 MFLX06450-08 MFLX06450-07 MFLX06450-09 MFLX06406-18 MFLX06406-76 MFLX06406-64 MFLX06406-12 MFLX06406-66 MFLX06406-72 MFLX06406-60 MFLX06406-74 MFLX06406-63 MFLX06406-62 MFLX06450-00 MFLX06450-02 MFLX06450-01 MFLX06450-15 MFLX06450-04 MFLX06450-14 MFLX06450-06 MFLX06450-17 MFLX06450-05
Masterflex® Transfer Tubing, Fluorinated Etyhlene Propylene (FEP), Avantor®
Tubing Transfer Tubing
Does not deteriorate with age.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application

Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Excellent for the transfer of extreme temperature or highly corrosive materials. Also good for high-purity work.
Characteristics: Chemically inert, nontoxic, heat resistant, with a low coefficient of friction. Doesn’t deteriorate with age. Tough, but bends even at -454 °F (-270 °C). Translucent.
Certifications: Resin is FDA-compliant (21 CFR 177.1550). Tubing is manufactured in ISO 9001-2008 approved facility.
Temperature range: -454 to 400 °F (-270 to 205 °C)
Sterilize: Autoclave, ethylene oxide, or use dry heat.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty