Would you like to log in again?
Metric Size Straight Barbed Reducing Fittings
Supplier: Scienceware
19561 19563 19559 19562 19558 19559
MFLX06458-21EA 584.34 USD InStock
MFLX06458-21 MFLX06458-10 MFLX06458-40 MFLX06458-50 MFLX06458-20 MFLX06458-60
Metric Size Straight Barbed Reducing Fittings
Fittings and Connectors
Ideal for low-pressure applications.
  • Quick and easy assembly without tools
  • Chemical resistant
Hose barb fittings make tubing connections quick and easy, requiring no tools for installation. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty