Would you like to log in again?
Lever-Action Drum Pumps with Nonmetallic-Body
Supplier: ACTION PUMP COMPANY
P490 R490ST 3007 P490S
MFLX06511-50EA 54.6 USD InStock
MFLX06511-50 MFLX06511-40 MFLX06511-60 MFLX06511-80
Lever-Action Drum Pumps with Nonmetallic-Body
Pumps Drum/Transfer Pumps
Economical pumps for use in cabinets or tight work spaces.
  • Fit 15, 30, and 55-gallon drums with 2" IPS bung openings
  • Designed for use in industrial, agricultural, labs, and automotive industries
Choose from center-level pump and lever-action pump model options.
Center-Lever Pump is self-priming and features an anti-drip spout to eliminate wasted fluid. This pump is ideal for oils, detergents, and other compatible industrial fluids.
Lever-Action Pumps feature a curved discharge spout for convenient hose extension. Choose from three models depending on your chemical needs. Model 06511-50 is ideal for water, alcohol, gasoline, diesel, kerosene, and lubricants. Model 06511-60 handles the same fluids, plus water-based chemicals, acids, and alkalis. Model 06511-80 handles the same fluids, plus strong acids and strong alkalis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty