Would you like to log in again?
Hand-Operated Piston Dispensing Drum Pump, Polypropylene
Supplier: ACTION PUMP COMPANY
6008 6009
MFLX06512-05EA 37.9 USD InStock
MFLX06512-05 MFLX06512-00
Hand-Operated Piston Dispensing Drum Pump, Polypropylene
Pumps Drum/Transfer Pumps
Economical piston pumps are ideal for a variety of fluids.
  • Adaptable to most pails and drums
Models 06512-00 and -05 have adjustable strokes to allow for dispensing different amounts fo fluid with just one pump. Model 06512-00 pumps 64, 32, 22 and 16 strokes per gallon, model 60512-05 pumps 32, 16, 11 and 8 strokes per gallon.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty