Would you like to log in again?
Masterflex® I/P® Long-Shaft Pump Head for Precision Tubing, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
77602-02 77601-62 7019-26 7019-25 77602-12 7019-00
MFLX07019-00EA 725 USD InStock
MFLX07019-00 MFLX77601-62 MFLX07019-25 MFLX07019-26 MFLX77602-02 MFLX77602-12
Masterflex® I/P® Long-Shaft Pump Head for Precision Tubing, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Ideal for fluid transfer applications requiring high accuracy and repeatability.
  • Designed and optimized for one tubing size
  • Three roller design for smooth flow
  • Choose from stainless steel (SS) or cold-rolled steel (CRS) rotor / roller for greater durability and chemical resistance
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
Standard heads are precision molded to accommodate a single size of tubing—use for common fluid transfer applications requiring accuracy and excellent repeatability. Available with clear polycarbonate (PC) housing with either cold-rolled steel (CRS) or stainless steel (SS) rotors; CRS offers economy, while SS offers better chemical resistance and durability.
Use with Masterflex® I/P® pumps requiring the long-shaft style pump head. These pump heads are not compatible with standard shaft drives.
Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications. Standard heads are compact and easy to integrate into your OEM applications. Call for more information on our comprehensive OEM services.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty