Would you like to log in again?
Masterflex® I/P® Standard Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
7019-53 7019-43 7019-31 7019-32 7019-21 7019-20
MFLX07019-43EA 1110 USD InStock
MFLX07019-43 MFLX07019-32 MFLX07019-21 MFLX07019-53 MFLX07019-31 MFLX07019-20
Masterflex® I/P® Standard Pump Heads, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Best accuracy and repeatability of any I/P pump head—ideal for dispensing and metering.
  • Each model is optimized for a single tube size to maximize performance and tubing life
  • Cost effective pump head is an excellent choice for dedicated applications
  • Stack two pump heads to double flow rate
The Standard pump head’s classic design ensures the most accurate and repeatable performance—important in dispensing applications. A 360-degree precision molded occlusion body ensures optimum accuracy and long tubing life.
Select from PC housing with CRS or SS rotor, or PPS housing with SS rotor. PC is economical and transparent, while PPS offers enhanced chemical resistance. SS rotors feature PTFE-sealed SS bearings for long life and reduced maintenance; CRS rotors have shielded bearings.
The design of the standard pump head enables the most precise dispensing accuracy of any MASTERFLEX pump head.
Delivery information: Includes single channel mounting hardware, replaceable tang boot for quiet operation and long service life, and tubing loading key. To mount two pump heads on a single drive, order dual channel mounting hardware 07019-96 under "Accessories".
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty