Would you like to log in again?
Air-Powered Polypropylene Pump Head Standard Centrifugal Pumps
Supplier: MARCH MANUFACTURING INC
0150-0120-0500 0130-0018-0700 0125-0088-0300
MFLX07022-80EA 774 USD InStock
MFLX07022-80 MFLX07022-85 MFLX07022-84
Air-Powered Polypropylene Pump Head Standard Centrifugal Pumps
Pumps Centrifugal Pumps
Air-operated pump is ideal for intrinsically safe areas.
  • Use pumps in chemical plants, organic compound transfer applications, and wastewater treatment centers
Order an air lubricator and an air filter/regulator under accessories to remove impurities in the air supply.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty