Would you like to log in again?
Flux Pumps Electric Drum Pump TEFC Motors
Supplier: FLUX PUMPS CORP
F458 120V
MFLX07076-02EA 1210 USD InStock
MFLX07076-02
Flux Pumps Electric Drum Pump TEFC Motors
Pumps Drum/transfer Pumps
Configurable to your requirements.
  • High-viscosity tube set handles fluids with viscosities to 50,000 cp!
Electric motors are ideal for difficult pumping situations. Splashproof, corrosion-resistant epoxy coating resists caustic fluids. Built-in overload protection switch has manual reset for safety.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty