Would you like to log in again?
Hand-Operated Piston Dispensing Drum Pump, PVC
Supplier: ACTION PUMP COMPANY
507 506 505 504
MFLX07079-65EA 146 USD InStock
MFLX07079-65 MFLX07079-55 MFLX07079-60 MFLX07079-50
Hand-Operated Piston Dispensing Drum Pump, PVC
Pumps Drum/transfer Pumps
Economical piston pumps are ideal for a variety of fluids.
  • Adjustable to most pails and drums
Models 07079-50 and -55 feature an angled spout to help prevent dripping. Models 07079-60 and -65 are made entirely of PVC, including the piston and seal, making them an ideal option for sanitary applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty