Would you like to log in again?
Guzzler® Portable Diaphragm Hand Pumps
Supplier: Guzzler
G500-H COLEP G2600 G400-H COLEP G450-H
MFLX07090-09EA 178 USD InStock
MFLX07090-09 MFLX07090-10 MFLX07090-11 MFLX07090-12
Guzzler® Portable Diaphragm Hand Pumps
Pumps Drum/Transfer Pumps
Low-cost, high-flow pumping solution.
  • Ideal for pumping shear-sensitive or particulate-laden fluids
  • Durable, portable pumps mount in any position
  • Available in cast aluminum or nonclogging Delrin® acetal plastic
  • 07090-09 has inlet and outlet on same side
Guzzler® hand pumps are designed for quick, easy fluid transfer.
Order service kits, noncollapsing wire-reinforced vinyl hoses, stainless steel hose clamps, and check valves separately in table.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty