Would you like to log in again?
Masterflex® L/S® PTFE-Diaphragm Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
7090-42 7090-62
MFLX07090-62EA 1070 USD InStock
MFLX07090-62 MFLX07090-42
Masterflex® L/S® PTFE-Diaphragm Pump Heads, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Compatible with most Masterflex® L/S® drives.
  • PTFE and glass wetted parts help maintain fluid purity
  • Ideal for pumping through filters or into pressurized systems
  • Check valve on inlet side of pump holds prime for up to two hours when pump is off
Note: Do not use these pump heads with Masterflex® drives requiring a long-shaft pump head.
Delivery information: Includes mounting tools, coupling hardware, and service kit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty