Would you like to log in again?
FMI Valveless Piston Pump Heads, Series Q
Supplier: FMI
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FMI353 / Q2CKY FMI364 / Q2SSY FMI353 / Q2CKY FMI205 / Q1CSC FMI217 / Q3CKC FMI352 / Q1CKC + (2) R412-2K FMI363 / Q1SSY FMI361 / Q1SKY FMI358 / Q1CKY FMI360 / Q2CSY Q2CSC FMI359 / Q1CSY FMI362 / Q2SKY FMI202 / Q0SSY FMI316 / Q0SKY FMI355 / Q2CKC FMI462 / Q3CSC
MFLX07117-26EA 534.5 USD InStock
MFLX07117-26 MFLX07104-70 MFLX07104-60 MFLX07104-72 MFLX07104-50 MFLX07117-28 MFLX07104-66 MFLX07104-56 MFLX07104-68 MFLX07104-58 MFLX07104-62 MFLX07104-52 MFLX07104-63 MFLX07104-53 MFLX07104-64 MFLX07104-54 MFLX07104-59
FMI Valveless Piston Pump Heads, Series Q
Pumps Pump Heads Piston Pump Heads
No valves to clog or wear out.
  • Build your own pump systems using interchangeable heads, drives, and accessories
  • Pump fluids with viscosities up to 500 cp
  • Low dead volume
These FMI Series Q valveless piston pump heads give you options to handle a wide range of fluids at pressures up to 100 psi. The interchangeable pump head modules provide numerous flow and material combinations for optimal versatility. Choose from a combination of ceramic, PVDF, carbon, and stainless steel wetted materials for an inert and corrosion resistant fluid path, ideal for your specific application. These materials offer broad fluid compatibility for dependable long-life operation.
To connect the stainless steel pump head module to 1/4" tubing please use adapter 07104-81. To connect to 3/8" ID tubing, please select adapter 07104-83. These are sold in quantities of one and one will normally need to have two to complete their system.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty