Would you like to log in again?
Gems LS-3 Series Level Switches
Supplier: Gems Sensors & Controls
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
142545 79990 M5600 171517 173250 43765 01701 142505 74780 171514 01702 116826
MFLX07187-20EA 277 USD InStock
MFLX07187-20 MFLX07188-40 MFLX07187-18 MFLX07187-08 MFLX07187-16 MFLX07187-06 MFLX07187-14 MFLX07187-04 MFLX07187-12 MFLX07188-35 MFLX07187-02 MFLX07187-10
Gems LS-3 Series Level Switches
Level Sensors
Install from the top of your tank—attach your own conduit to protect wiring.
  • Get longer life and more durability with hermetically sealed switches
All models accept 120/240 VAC and operate as NO or NC switches except where noted. Switches include 24" leads (except 07187-12 which has 72" leads). If your level control system exceeds the current rating in the table, use a controller with your switch.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty