Would you like to log in again?
Masterflex® L/S® Standard Digital Drives, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
7522-30 7522-28 7522-20 7522-20 7522-28 7522-20 7522-20 7522-30 7522-20 7522-20 7522-20 7522-28 7522-38 7522-38 7522-28 7522-30 7522-28 7522-20 7522-30 7522-38 7522-28 7522-38 7522-30 7522-28 7522-38 7522-38 7522-20 7522-30 7522-28 7522-38 7522-38 7522-30 7522-38 7522-30 7522-30 7522-28
MFLX07522-30-INCS 0 USD InStock
MFLX07522-30-IN MFLX07522-20 MFLX07522-30-CH MFLX07522-20-UK MFLX07522-28-IL MFLX07522-38-EU MFLX07522-30-IT MFLX07522-28 MFLX07522-28-IT MFLX07522-38-AU MFLX07522-20-IL MFLX07522-30-EU MFLX07522-28-CH MFLX07522-28-IN MFLX07522-20-IN MFLX07522-20-CH MFLX07522-28-AU MFLX07522-30-UK MFLX07522-20-IT MFLX07522-38-IL MFLX07522-28-EU MFLX07522-20-EU MFLX07522-38-IN MFLX07522-38-CH MFLX07522-38-IT MFLX07522-20-AU MFLX07522-30-IL MFLX07522-30 MFLX07522-30-DK MFLX07522-38-UK MFLX07522-38 MFLX07522-28-DK MFLX07522-20-DK MFLX07522-30-AU MFLX07522-38-DK MFLX07522-28-UK
Masterflex® L/S® Standard Digital Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Designed with maintenance-free high-accuracy motor and a graphical interface—makes setup and operation easier than ever!
  • Flow rate 0.001/0.006 to 566/3400 ml/min with L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating modes—continuous run, timed dispense, copy dispense, or volume dispense
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle)
  • Antidrip function ensures dispensing accuracy
  • Brushless, maintenance-free motor offers ±0.1% speed control accuracy with a 6000 to 1 turndown ratio
These precision drives offer analog remote control options and simple programming—ideal for automated process applications. Drives feature a programmable dispense interval that lets you set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—drive stores one calibration value per tubing size.
Program the drive via the simple membrane keypad; keypad lock/unlock feature protects settings. Motor is reversible so you can purge tubing before or after pumping and pump fluid in either direction.
Ordering information: Each model includes a country specific plug/cord set; choose the catalog number with desired country plug type. All units include an IEC 320 socket with a detachable standard US three prong power cable. See country plug description https://us.vwr.com/cms/tech-article-ac-power-plugs-sockets-universal-pattern-guide.
Delivery information: Includes pump drive and power adapter with 1.8 m (6 ft.) line cord. These models are shipped with country specific plug/cord set; please order the specific catalog number with the desired plug type.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty