Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Masterflex® L/S® Digital Miniflex® Pump Systems, Avantor®

Supplier: Avantor Fluid Handling

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Masterflex®
7525-20 7525-40 7525-20 7525-40 7525-20 7525-40 7525-20 7525-40 7525-20 7525-40 7525-40 7525-20 7525-20 7525-40 7525-40 7525-40 07525-20 7525-20
MFLX07525-20-DKCS 0 USD InStock
MFLX07525-20-DK MFLX07525-40-IL MFLX07525-40-IN MFLX07525-40-CH MFLX07525-20-IL MFLX07525-40-DK MFLX07525-20-CH MFLX07525-20-IN MFLX07525-20-EU MFLX07525-40-IT MFLX07525-20-IT MFLX07525-20-UK MFLX07525-40-EU MFLX07525-40-AU MFLX07525-20-AU MFLX07525-40-UK MFLX07525-40 MFLX07525-20
Masterflex® L/S® Digital Miniflex® Pump Systems, Avantor®
Pumps Peristaltic Pumps
Make low-flow pumping easy!

  • Flow range 0.004 to 315 ml/min—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode
  • Remote control available using analog communication—voltage, current and contact closure
  • Integrated “open-head” sensor halts operation when pump head is opened
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Automatic tubing retention makes tube loading quick and easy
  • More than 20% smaller than competitive units—compact size and light weight make it easy to move and share between labs

This compact, stackable pump features a durable ABS housing that is easy to wipe down and keep clean. The Miniflex pump head features a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life.

Note:Dual-channel model must have same size tubing loaded in both channels to ensure tubing retention.

Delivery information: Includes l/S Miniflex pump head and universal voltage (90 to 260 VAC) benchtop power supply with IEC320/CEE22 socket. Pump is shipped with cord/plug assembly specific to country of destination.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty