Would you like to log in again?
Masterflex® L/S® Analog Variable-Speed Console Drives, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
7555-15 7555-15 7555-10 7555-05 7555-15 7555-05 7555-15 7555-15 7555-15 7555-05 7555-05 7555-05 7555-10 7555-15 7555-15 7555-00 7555-05 7555-05 7555-05 7555-15 7555-05
MFLX07555-15-DKCS 0 USD InStock
MFLX07555-15-DK MFLX07555-15-IL MFLX07555-05-UK MFLX07555-05-AU MFLX07555-15-CH MFLX07555-15-IN MFLX07555-15 MFLX07555-05 MFLX07555-00 MFLX07555-05-EU MFLX07555-10-USS MFLX07555-10 MFLX07555-05-IT MFLX07555-15-AU MFLX07555-05-DK MFLX07555-15-UK MFLX07555-05-IN MFLX07555-05-CH MFLX07555-05-IL MFLX07555-15-EU MFLX07555-15-IT
Masterflex® L/S® Analog Variable-Speed Console Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Compact and cost effective for general fluid transfer applications.
  • Flow rate 0.42 to 2900 ml/min with L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Simple operation with single-turn speed control
  • Separate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power on
  • Reversible motor lets you easily prime/purge tubing and pump in either direction
  • Remote control of start/stop via DB9 connector on back of drive
Drives accept nine different pump head types: Standard, Easy-Load®, Easy-Load II, Easy-Load 3, Multichannel, High-Performance, Cartridge, PTFE tubing, and PTFE-diaphragm. Drives 07555-00 and -05 are not recommended for use with the PTFE-tubing and PTFE-diaphragm pump heads.
Ordering information: Each model includes a country specific plug/cord set; choose the catalog number with desired country plug type. All units include an IEC 320 socket with a detachable standard US three prong power cable. See country plug description https://us.vwr.com/cms/tech-article-ac-power-plugs-sockets-universal-pattern-guide.
Delivery information: Includes pump drive and power adapter with 1.8 m (6 ft.) line cord. These models are shipped with country specific plug/cord set; please order the specific catalog number with the desired plug type.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty