Would you like to log in again?
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Peroxide-Cured Silicone, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
SC0095/ 2 STOP SC0105 SC0101 SC0104 SC0103/ 2 STOP SC0099 SC0093/2 STOP SC0092/2 STOP SC0097 116-0497-04 SC0094/ 2 STOP SC0098 SC0102/2 STOP SC0100 SC0096/2 STOP
MFLX07616-38EA 102 USD InStock
MFLX07616-38 MFLX07616-48 MFLX07616-26 MFLX07616-36 MFLX07616-46 MFLX07616-24 MFLX07616-28 MFLX07616-30 MFLX07616-40 MFLX07616-12 MFLX07616-34 MFLX07616-44 MFLX07616-22 MFLX07616-32 MFLX07616-42
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Peroxide-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Long service life - outstanding fatigue resistance due to flexing!
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Appearance: Opaque
Temperature: -70 to 52 °C
Life at 100 rpm: 1000 hrs
The first of a series of two-stop extension tubing options is below under Accessories. See all of our two-stop extension tubing formulations or contact our Application Specialists for personal assistance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty