Would you like to log in again?
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Peroxide-Cured Silicone, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
SC0116/3 STOP SC0108/3 STOP SC0110/3 STOP SC0113 SC0115 SC0109/3 STOP SC0114 SC0112 SC0106/ 3 STOP SC0117/3 STOP SCO107/3 STOP SC0111 3-STP PRXD SLCN 0.51 SC0118 SC0119
MFLX07624-40CS 97 USD InStock
MFLX07624-40 MFLX07624-42 MFLX07624-30 MFLX07624-18 MFLX07624-22 MFLX07624-44 MFLX07624-32 MFLX07624-24 MFLX07624-46 MFLX07624-34 MFLX07624-26 MFLX07624-48 MFLX07624-36 MFLX07624-28 MFLX07624-38
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Peroxide-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attached tubing stops give you two sections—when one section of tubing fatigues, a fresh section is ready to use. The stops on the three-stop tubing are color-coded according to tubing ID. Use three-stop tubing with any of the Ismatec® minicartridge pumps and pump heads. Minicartridges withstand pressures up to 22 psi.
The peroxide-cured silicone tubing features an ultra-smooth inner bore, excellent biocompatibility for sensitive applications, non-cytotoxic, non-pyrogenic, and non-hemolytic, low protein binding, heat-sealable, bondable, sterile-weldable, and custom moldable for assemblies. It is ideal for pharmaceutical, biotech, and general-purpose lab applications, including sterile filling and processing, cell harvest and media process systems, filtration, fermentation, cell and tissue culture transport. It features a clear appearance, has a temperature range of –66 to 135 °C, and has a pumping life of approximately 1000 hours at 100 rpm.
The overall length of peroxide-cured silicone 07624-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty