Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Rubberfab Nylon Clamps for Sanitary Fittings
Supplier: Rubberfab
13MHHM-NGW-150 13MHHM-NGW-050/075
MFLX12805-03EA 35.7 USD InStock
MFLX12805-03 MFLX12805-04
Rubberfab Nylon Clamps for Sanitary Fittings
Clamps Tubing and Sanitary Clamps
Collared clamp helps prevent unintentional opening.
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty