Would you like to log in again?
Value Plastics Luer Fittings and Adapters
Supplier: Value Plastics
MTLL430-6005 BSMLRL-6005 LP4-9 FSLLR-6005 MTLSP-1 LP4-6005 LE87-6005 FTLLB220-1 FTLLP-6005 SFTL-6005 MTLS220-1 MLRL055-1 ABSML-6005-1 (1,000PK) FTLL430-6005 MTLS230-1 FTLL210-6005 MTLS230-5 FTLL004-6005 MTLL240-6005 FTLL240-1 MLRLB210-6005 MTLL025-1 RMLLR-6005 ABSML-6005-1 (25PK) FTLL230-6005 MTLL230-6005 MTLL250-6005 MTLLP-6005 MTLL007-9
MFLX15955-93EA 65.2 USD InStock
MFLX15955-93 MFLX30651-72 MFLX15955-79 MFLX15955-78 MFLX15955-98 MFLX15955-97 MFLX15973-20 MFLX15955-96 MFLX15955-95 MFLX15973-12 MFLX15973-11 MFLX15973-17 MFLX15973-14 MFLX15973-15 MFLX15973-18 MFLX15955-82 MFLX15955-81 MFLX15955-87 MFLX15955-84 MFLX15955-83 MFLX15973-45BLK MFLX30495-72 MFLX15973-45 MFLX15956-09 MFLX15956-08 MFLX15956-07 MFLX15973-25 MFLX15973-26 MFLX15973-29
Value Plastics Luer Fittings and Adapters
Fittings and Connectors
Premium luer fittings for critical applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • Full material traceability on all items
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Manufactured in compliance with ISO 594-1 standards, these fittigns are inspected to be free from contaminants, and are fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1/16" to 1/4", with a variety of connection options. All fittings are sold in packs of 100.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty