Would you like to log in again?
PendoTECH® Single-Use Pressure Sensors for Pressure Monitor and Transmitter Systems
Supplier: PendoTech
PendoTECH®
PREPS-N-075 PREPS-N-5-038 PRESS-S-000 PRESS-N-050 PREPS-N-100 PRESS-N-100 PREPS-N-050 PREPS-N-1-1 PREPS-N-5-050 PREPS-N-1-100 PREPS-N-012 PRESS-N-038 PREPS-N-000 PRESS-N-025 PREPS-N-038 PRESS-N-075 PREPS-N-025 PREPS-N-5-5
MFLX19406-56EA 132 USD InStock
MFLX19406-56 MFLX19406-23 MFLX19406-55 MFLX19406-22 MFLX19406-25 MFLX19406-24 MFLX19406-27 MFLX19406-29 MFLX19406-28 MFLX19406-30 MFLX19406-52 MFLX19406-51 MFLX19406-32 MFLX19406-54 MFLX19406-21 MFLX19406-31 MFLX19406-53 MFLX19406-20
PendoTECH® Single-Use Pressure Sensors for Pressure Monitor and Transmitter Systems
Fluid Sensors
Single-use solution to monitor process pressure.
  • Ideal for applications such as tangential flow filtration, cross flow filtration, depth filtration, bioreactor monitoring, and chromatography
  • Sensors priced for single-use systems, but may be reused
Sensors measure static and dynamic process pressure and are perfect components for disposable systems requiring sterilization, but also minimal downtime. Sensors feature an integral flow sensor with no dead legs and no fluid path obstructions. All pressure sensors are manufactured in a FDA compliant facility, in an ISO Class 8 clean room. Sensors are made from USP Class VI-compliant polycarbonate or caustic-resistant polysulfone. Both materials are gamma stable. Sensors are 100% tested for accuracy and leaks during the manufacturing process. Connect sensor to PressureMAT unit with 12-ft (3.6-m) reusable cable (included). Sensors are designed for use up to a maximum pressure of 5.3 bar. Available in either process–scale or small–scale sizes.
Process-Scale Sensors connect directly as a barbed fitting to the tubing line and are available in 1⁄4", 3⁄8", 1⁄2", 3⁄4", or 1" hose barb or 3/4" or 1" sanitary flange.
Small-Scale Sensors are available with luer connections and may connect directly to the fluid line with luer adapters or indirectly by using a 1⁄4" or 3⁄8" barbed adapter tee. Both connection setups have been tested for accuracy.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty