Would you like to log in again?
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
40RXPX-150 40MPE-150 40RXPX-200 40RXPX-100 40RXPX-400 40MPU-250 40MPE-400 42MPU-.750 42MPSFY-075 40MPSFY-250 40MPG-250 40MPG-200 40MPG-100 40MPU-100 40RXPX-300 42RXPX-050 40MPE-250 40RXPX-W-200 40MPE-100 42RXPX-075 40MPU-400 42RXPX-050 40MPSFY-200 40RXPX-600 40MPG-150 42MPE-075 40MPU-200 42MPG-075 40MPSFY-150 40MPSFY-100 40RXPX-F-400 40MPSFY-400 40MPU-150 40MPE-200 40RXPX-250
MFLX16011-66EA 25.6 USD InStock
MFLX16011-66 MFLX00404-MO MFLX00162-NJ MFLX00364-YQ MFLX30548-04 MFLX30548-48 MFLX30548-26 MFLX30548-02 MFLX30548-68 MFLX00158-NC MFLX30548-46 MFLX30548-24 MFLX41001-77 MFLX30548-08 MFLX30548-06 MFLX30548-28 MFLX30548-40 MFLX30548-62 MFLX30548-84 MFLX30548-82 MFLX30548-60 MFLX30548-88 MFLX30548-00 MFLX30548-22 MFLX30548-44 MFLX30548-66 MFLX30548-64 MFLX30548-20 MFLX30548-86 MFLX30548-42 MFLX29590-82 MFLX30548-80 MFLX29590-81 MFLX29590-80 MFLX29590-83
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Seals Gaskets
Ensure a tight seal.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class VI and cytotoxicity criteria.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from –58 to 446 °F (–50 to 230 °C).
Viton offers excellent chemical resistance, especially to many acids and alkalis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty