Would you like to log in again?
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Elbow, HDPE, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
L4-4HDPE(10 PACK) L2-5HDPE(10 PACK) L8-8HDPE(10 PACK) L8-4HDPE(10 PACK) ELB-1836B-10 L4-8HDPE(10 PACK) L2-4HDPE 100 PK L16-16HDPE 10PK L8-6HDPE(10 PACK) L6-6HDPE(10 PACK) L4-6HDPE(100 PACK) L4-2HDPE(100 PACK) L4-3HDPE(10 PACK) L12-12HDPE 10PK L2-6HDPE 10 PK L4-6HDPE(10 PACK) L6-8HDPE(10 PACK) L8-10HDPE(10 PACK) L6-4HDPE(100 PACK) L6-4HDPE(10 PACK) L4-2HDPE(10 PACK) L8-12HDPE 10PK L2-2HDPE(10PK) L2-4HDPE 10 PK
MFLX30622-92EA 25.3 USD InStock
MFLX30622-92 MFLX31518-77 MFLX30622-82 MFLX30626-88 MFLX30627-02 MFLX30626-98 MFLX30622-96 MFLX30623-10 MFLX30623-00 MFLX30401-29 MFLX30622-98 MFLX30401-06 MFLX30623-12 MFLX30622-88 MFLX30401-28 MFLX30623-02 MFLX30623-14 MFLX30401-08 MFLX30623-04 MFLX30623-06 MFLX30623-08 MFLX30401-31 MFLX30401-04 MFLX31702-30
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Elbow, HDPE, Avantor®
Fittings and Connectors
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed elbow fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty