Would you like to log in again?
Masterflex® Sanitary Caps, 316SS
Supplier: Avantor Fluid Handling
Masterflex®
16AMP-200-316L-HDL 16AMP-075-316 16AMP-250-316 16AMP-150-316 16AMP-300-316L-HDL 16AMP-250-316L-HDL 16AMP-200-316 16AMP-300-316 16AMP-100/150-316L-HDL
MFLX30510-81EA 18.9 USD InStock
MFLX30510-81 MFLX30510-92 MFLX30510-93 MFLX30510-82 MFLX30510-83 MFLX30510-84 MFLX30510-90 MFLX30510-91 MFLX30510-80
Masterflex® Sanitary Caps, 316SS
Fittings and Connectors
High-quality tri-clamp to hose barb fittings in 316 stainless steel for sanitary applications.
  • Constructed from 316 stainless steel for excellent corrosion resistance and durability
  • Designed to meet sanitary standards, ensuring hygienic fluid transfer in critical applications
  • Secure tri-clamp design allows for reliable attachment to various equipment and systems
  • Tailored hose barb shape and size options to accommodate a wide range of hose sizes
  • Precision engineered and designed for use with tubing clamps to ensure tight seals for leak-free connections, maintaining product integrity
  • Suitable for various industries including food and beverages, pharmaceuticals, and more
  • Capable of handling demanding operating conditions with resistance to high temperatures and pressures
  • Designed to work seamlessly with existing equipment, enhancing overall system compatibility
Elevate your fluid transfer systems with these premier sanitary tri-clamp to hose barb fittings in 316 stainless steel. Engineered for excellence, these fittings combine top-grade materials with hygienic design, ensuring unparalleled durability, corrosion resistance, and leak-free performance. Their versatile compatibility with various hose types and sizes, seamless tri-clamp attachment, and easy maintenance make installation a breeze, while adherence to strict industry standards guarantees a contamination-free transfer for industries ranging from food and beverage to pharmaceuticals. Trust in these fittings to enhance operational efficiency, promote system integrity, and set new benchmarks for fluid management excellence.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty