Would you like to log in again?
Masterflex® Platinum-cured Silicone Screen Gaskets, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
40RXPXS150-60SS 40RXPXS250-100SS 40RXPXS100-10SS 42RXPXS075-60SS 42RXPXS050-100SS 40RXPXS150-40SS 40RXPXS100-60SS 40RXPXS200-100SS 40RXPXS100-40SS 40RXPXS150-100SS 40RXPXS150-10SS 40RXPXS100-200SS 42RXPXS050-20SS 40RXPXS150-20SS 40RXPXS200-40SS 42RXPXS050-60SS 42RXPXS075-20SS 40RXPXS100-20SS 42RXPXS050-40SS 40RXPXS200-60SS 40RXPXS300-10SS 42RXPXS075-40SS 42RXPXS075-100SS 40RXPXS200-325SS 40RXPXS100-100SS 40RXPXS200-10SS 40RXPXS150-200SS 40RXPXS200-20SS
MFLX00383-POEA 91 USD InStock
MFLX00383-PO MFLX41000-50 MFLX00253-BM MFLX30523-08 MFLX30523-46 MFLX30523-24 MFLX30523-68 MFLX30523-66 MFLX30523-88 MFLX30523-44 MFLX30523-22 MFLX30523-06 MFLX30523-28 MFLX30523-04 MFLX30523-26 MFLX30523-48 MFLX00269-AW MFLX00431-JA MFLX30523-60 MFLX30523-82 MFLX30523-80 MFLX30523-64 MFLX30523-86 MFLX30523-20 MFLX30523-42 MFLX30523-62 MFLX30523-40 MFLX30523-84
Masterflex® Platinum-cured Silicone Screen Gaskets, Avantor®
Seals Gaskets
Protect your pump, valves and other components in fluid line.
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derived ingredient free. Each gasket sold individually.
Platinum-cured silicone gaskets are the ideal choice for pharmaceutical, high-purity water, injectable, food, and beverage applications. Gaskets will resist product adhesion, and are tasteless, odorless and do not contain plasticizers. Use silicone gaskets at temperatures from –58 to 446 °F (–50 to 230 °C).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty