Would you like to log in again?
Plastomatic Sanitary Plastic Ball Valves
Supplier: Plast-O-Matic
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MBV075EPSC-PF MBV100EPSC-PF MBV075EPSC-PP MBV100EPSC-PP MBV150EPSC-PP MBV100EPSCM-PP MBV150EPSC-PF MBV200EPSC-PF
MFLX30532-11EA 806 USD InStock
MFLX30532-11 MFLX30532-01 MFLX30532-15 MFLX30532-03 MFLX30532-17 MFLX30532-05 MFLX30532-19 MFLX30532-07
Plastomatic Sanitary Plastic Ball Valves
Valves Ball Valves
Precision machined and tested to ensure leak-free connections.
  • Standard Tri-Clamp connections are easy to plumb into sanitary fluid paths
  • Smooth flow path minimizes turbulence and pressure loss
  • Valve design permits full flow and pressure in either direction
These industry standard valves feature PTFE bearings and seats to virtually eliminate friction and minimize wear. Fully concentric polished ball further ensures leak-free performance. Available in polyproplyene (PP) or PVDF body material with EPDM seals.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty