Would you like to log in again?
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, EPDM, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
42MPES050-20SS 40MPES150-10SS 40MPES200-100SS 42MPES075-60SS 40MPES100-20SS 40MPES150-40SS 40MPES200-60SS 42MPES050-100SS 40MPES150-20SS 42MPES050-60SS 40MPES150-100SS 42MPES050-40SS 40MPES100-60SS 40MPES150-60SS 42MPES075-20SS 40MPES200-20SS 40MPES100-100SS 42MPES075-40SS 40MPES100-40SS 40MPES100-10SS 40MPES200-40SS 40MPES200-10SS 42MPES075-100SS
MFLX30537-80EA 65 USD InStock
MFLX30537-80 MFLX30537-26 MFLX30537-04 MFLX30537-48 MFLX30537-24 MFLX30537-46 MFLX30537-68 MFLX30537-08 MFLX30537-06 MFLX30537-28 MFLX30537-62 MFLX30537-40 MFLX30537-84 MFLX30537-60 MFLX30537-82 MFLX30537-88 MFLX30537-66 MFLX30537-22 MFLX30537-44 MFLX30537-20 MFLX30537-86 MFLX30537-64 MFLX30537-42
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, EPDM, Avantor®
Seals Gaskets
Protect your pump, valves and other components in fluid line.
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derived ingredient free. Each gasket sold individually.
EPDM gaskets offer an economical solution to transfer water, acetones, and ketones. Gaskets are ideal in chemical process applications; temperature range is –65 to 212 °F (–54 to 100 °C).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty