Would you like to log in again?
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, PTFE, Avantor®
Supplier: Avantor Fluid Handling
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Masterflex®
42MPGS050-20SS 40MPGS150-100SS 42MPGS050-100&10SS 40MPGS200-60SS 40MPGS150-20SS 40MPGS200-20SS 40MPGS150-10SS 40MPGS150-40SS 40MPGS100-40SS 42MPGS050-60SS 40MPGS100-20SS 40MPGS100-60SS 40MPGS100-10SS 42MPGS075-60SS 40MPGS200-40SS 40MPGS100-100&10SS 40MPGS200-100&10SS 40MPGS150-60SS 42MPGS050-40SS 40MPGS200-10SS
MFLX30538-80EA 188 USD InStock
MFLX30538-80 MFLX30538-60 MFLX61030-13 MFLX30538-08 MFLX30538-26 MFLX30538-04 MFLX30538-48 MFLX30538-28 MFLX30538-06 MFLX30538-44 MFLX30538-88 MFLX30538-66 MFLX30538-68 MFLX30538-46 MFLX30538-24 MFLX30538-84 MFLX30538-40 MFLX30538-20 MFLX30538-86 MFLX30538-64
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, PTFE, Avantor®
Seals Gaskets
Protect your pump, valves and other components in fluid line.
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derived ingredient free. Each gasket sold individually.
PTFE gaskets offer the best chemical compatibility and have exceptional resistance to high temperatures, corrosion, and stresscracking. PTFE gaskets will meet the highest demands in pharmaceutical, food and chemical applications. Use gaskets at temperatures from –450 to 550 °F (–268 to 288 °C).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty