Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
Masterflex® Tuf-Steel® Sanitary Screen Tri-Clamp Gaskets, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
40MPGTSS150-60SS 40MPGTSS200-20SS 40MPGTSS200-60SS 40MPGTSS200-40SS 40MPGTSS100-40SS 40MPGTSS100-100&10SS 42MPGTSS050-20SS 40MPGTSS150-20SS 42MPGTSS050-40SS 40MPGTSS200-100&10SS 40MPGTSS200-10SS 40MPGTSS150-40SS 40MPGTSS150-100&10SS
MFLX30539-08EA 196 USD InStock
MFLX30539-08 MFLX30539-48 MFLX30539-26 MFLX30539-28 MFLX30539-66 MFLX30539-44 MFLX30539-88 MFLX30539-68 MFLX30539-46 MFLX30539-84 MFLX30539-40 MFLX30539-20 MFLX30539-86
Masterflex® Tuf-Steel® Sanitary Screen Tri-Clamp Gaskets, Avantor®
Seals Gaskets
Protect your pump, valves and other components in fluid line.
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derived ingredient free. Each gasket sold individually.
Tuf-Steel gaskets are made from a unique blend of PTFE and 316L SS. This allows for repeated SIP cleaning without creeping and minimal heat expansion. Tuf-Steel gaskets have excellent chemical resistance and completely stop leaks when torqued correctly at 50 in-lbs. Use Tuf-Steel gaskets from –20 to 450 °F (–29 to 232 °C).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty