Would you like to log in again?
Masterflex® Smart Gasket® Sanitary Clamps, Stainless Steel, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
CL-TH-050/075-1 CL-TH-075-2 CL-TH-200-3-GLD CL-TH-050/075-1 CL-TH-100/150-2 CL-TH-200-4 CL-TH-200-2 CL-TH-300-1-GLD CL-TH-100/150-1 CL-TH-050/075IN-3 CL-TH-100/150-4 CL-TH-050/075-2 CL-TH-100/150-1 CL-TH-100/150-3 CL-TH-200-1 CL-TH-100/150-3 CL-TH-100/150-2 CL-TH-200-3
MFLX41001-54EA 454 USD InStock
MFLX41001-54 MFLX30544-11 MFLX30545-15 MFLX30544-15 MFLX30542-03 MFLX30543-15 MFLX30543-03 MFLX30542-11 MFLX30543-11 MFLX30542-19 MFLX30542-07 MFLX30543-19 MFLX30543-07 MFLX30542-15 MFLX30545-19 MFLX30544-07 MFLX30544-19 MFLX41001-49
Masterflex® Smart Gasket® Sanitary Clamps, Stainless Steel, Avantor®
Seals Gaskets
Create a secure connection with your Masterflex® Smart Gasket® sanitary gaskets.
  • Constructed of durable stainless steel (SS)
The Smart Gasket clamps are required with the gaskets because they have slots in them that allow for the different number of poarts. The clamps are specific to the number of ports on the gasket.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty