Would you like to log in again?
Smiths Medical® Manifold Fittings, Female Luer Locks
Supplier: Smiths Medical
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
B14345FMH (50PK) B14312FMH (50PK) B14314FMH (50PK) B14343FMH (50PK)
MFLX30600-43BLKEA 307 USD InStock
MFLX30600-43BLK MFLX30600-44BLK MFLX30600-42BLK MFLX30600-41BLK
Smiths Medical® Manifold Fittings, Female Luer Locks
Valves Ball Valves
Regulate flow without any leaking.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty