Would you like to log in again?
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, HDPE, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
A6-6HDPE(10 PACK) A8-4HDPE(10 PACK) A2-2HDPE(10 PACK) A6-5HDPE(100 PACK) A2-3HDPE-MF(10PK) A4-8HDPE(10 PACK) A4-4HDPE-MF(10PK) A4-8HDPE(100 PACK) A12-8HDPE(10 PACK) A6-5HDPE(10 PACK) A8-6HDPE(100 PACK) A8-6HDPE(10 PACK) MHB-1236B-10 A2-5HDPE 10PK A4-6HDPE(10 PACK) A2-6HDPE (100 PACK) MHB-1436B-10 A12-6HDPE(10 PACK) A4-5HDPE(10 PACK) A2-2.5HDPE 10PK A4-6HDPE(100 PACK) A8-8HDPE-MF(10PK) P3MCB2 A6-4HDPE(100 PACK) A8-10HDPE-MF(10PK) A6-4HDPE(10 PACK) A12-12HDPE(10 PACK) A2-4HDPE (100 PACK) A6-8HDPE(100 PACK) A4-4HDPE(100 PACK) A8-8HDPE(10 PACK) MHB-1836B-10 A8-12HDPE-MF(10PK) A16-16HDPE(10PK) A8-12HDPE(10 PACK) A8-10HDPE(10 PACK) MHB-1836B-100 A-4-2HDPE(10 PACK) A8-8HDPE(100 PACK) A2-6HDPE(10 PACK) A6-6HDPE A12-10HDPE(10 PACK) A2-5HDPE 100PK A6-6HDPE(100 PACK) P3MCB4 A4-6HDPE-MF(10PK) A8-4HDPE(100 PACK) P3MCB4 A2-2HDPE-MF(10PK) A6-8HDPE(10 PACK) A2-2HDPE(100 PACK) A4-4HDPE(10 PACK) P3MCB2 A12-4HDPE(10 PACK) A8-12HDPE(100 PACK) A4-2.5HDPE(10 PACK) A12-16HDPE(10 PACK) A2-4HDPE(10 PACK) A-4-2HDPE (100 PACK)
MFLX31517-21EA 21.1 USD InStock
MFLX31517-21 MFLX30625-42 MFLX30625-46 MFLX06444-16 MFLX06444-18 MFLX30621-48 MFLX30621-46 MFLX30621-42 MFLX30615-05 MFLX30615-09 MFLX30625-52 MFLX30625-54 MFLX30625-56 MFLX30625-58 MFLX31517-18 MFLX30621-08 MFLX30621-16 MFLX30621-58 MFLX30621-14 MFLX30615-13 MFLX30621-56 MFLX30621-12 MFLX30621-54 MFLX30621-52 MFLX30615-17 MFLX30401-72 MFLX31204-22 MFLX30401-74 MFLX31204-20 MFLX30401-73 MFLX30625-62 MFLX30625-64 MFLX30625-26 MFLX30621-18 MFLX30621-26 MFLX31702-08 MFLX31702-04 MFLX30621-66 MFLX30621-20 MFLX30615-21 MFLX30621-64 MFLX30621-62 MFLX30615-25 MFLX30615-29 MFLX30625-73 MFLX30625-75 MFLX30625-31 MFLX30625-77 MFLX30625-35 MFLX31702-18 MFLX30621-38 MFLX30621-35 MFLX30621-77 MFLX30621-75 MFLX30621-31 MFLX30621-73 MFLX30621-70 MFLX31701-82 MFLX30625-70
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, HDPE, Avantor®
Fittings and Connectors
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty