Would you like to log in again?
Overlook Industries OneShot Single-Use Filling Nozzles
Supplier: Overlook Industries
Masterflex®
OVCP-1/8-1-D OVCP-3/4-1-D OVCP-1/2-1-D OVCP-1/32-1-D OVCP-1/4-1-D OVCP-3/8-1-D OVCP-5/16-1-D OVCP-3/16-1-D OVCP-1/16-1-D
MFLX30619-04EA 243 USD InStock
MFLX30619-04 MFLX30619-03 MFLX30619-06 MFLX30619-05 MFLX30619-02 MFLX30619-01 MFLX30619-08 MFLX30619-07 MFLX30619-09
Overlook Industries OneShot Single-Use Filling Nozzles
Pumps Peristaltic Pumps
Reduce filler line downtime and risk of cross-contamination.
  • Lower costs for cleaning, sterilization, and validation
  • Provide enhanced flexibility in batch scheduling and product development
  • Dimensional tolerances of +/-0.05% for assured repeatability
These nozzles combine the ease of single-use plastic components with the accuracy and reliability of stainless steel filler needles. Tight dimensional tolerances help to ensure precision in your dispensing applications. Nozzles feature a 316L stainless steel needle and a polycarbonate base with hose barb adapter. Materials meet USDA and USP Class VI requirements, and is FDA compliant. All filling nozzles are sterilizable by ethylene oxide, autoclaving, or gamma irradiation (note: polycarbonate will turn a greenish-yellow hue when gamma irradiated).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty