Would you like to log in again?
Masterflex® Union Fittings, Hose Barb, Elbow, Stainless Steel, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
L0-4SS L0-5SS L0-8SS L0-3SS L0-10SS L0-2SS L0-12SS L0-6SS
MFLX30620-15EA 69.3 USD InStock
MFLX30620-15 MFLX30620-14 MFLX30620-13 MFLX30620-20 MFLX30620-19 MFLX30620-18 MFLX30620-17 MFLX30620-16
Masterflex® Union Fittings, Hose Barb, Elbow, Stainless Steel, Avantor®
Fittings and Connectors
Quick and easy assembly without tools.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty